Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256658.1 | NP_001243587.1 | 4 | 468 | ||
NM_001256659.1 | NP_001243588.1 | 4 | 424 | |||
NM_001256660.1 | NP_001243589.1 | 4 | 464 | |||
NM_001256661.1 | NP_001243590.1 | 4 | 439 | |||
NM_001256662.1 | NP_001243591.1 | 3 | 186 | |||
NM_003598.1 | NP_003589.1 | 4 | 424 | |||
XM_005259334.3 | XP_005259391.1 | 3 | 696 | |||
XM_006723424.1 | XP_006723487.1 | 3 | 696 | |||
XM_006723428.2 | XP_006723491.1 | 3 | 696 | |||
XM_011527398.1 | XP_011525700.1 | 3 | 696 | |||
XM_011527399.2 | XP_011525701.1 | 4 | 419 | |||
XM_011527400.2 | XP_011525702.1 | 4 | 389 | |||
XM_011527401.1 | XP_011525703.1 | 4 | 362 | |||
XM_011527402.2 | XP_011525704.1 | 4 | 364 | |||
XM_011527403.1 | XP_011525705.1 | 4 | 400 | |||
XM_011527404.1 | XP_011525706.1 | 3 | 696 | |||
XM_011527405.2 | XP_011525707.1 | 3 | 696 | |||
XM_011527406.2 | XP_011525708.1 | 3 | 696 | |||
GenBank mRNA | AK093236.1 | - | 3 | 696 | ||
AK290736.1 | - | 4 | 421 | |||
AK300241.1 | - | 3 | 170 | |||
AL832240.1 | - | 4 | 389 | |||
BC007556.1 | - | 4 | 424 | 2967735 | ||
BC018803.1 | - | 4 | 439 | |||
BC051301.1 | - | 4 | 418 | 4843913 | ||
X94440.1 | - | 3 | 240 |