Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201272.1 | NP_958429.1 | 16 | 2050 | ||
GenBank mRNA | AY499658.1 | - | 16 | 2050 |