Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tanc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002854.1 | NP_001002854.1 | 26 | 5599 | ||
XM_006234204.1 | XP_006234266.1 | 25 | 5518 | |||
XM_006234205.2 | XP_006234267.1 | 24 | 7754 | |||
XM_006234207.2 | XP_006234269.1 | 21 | 5716 | |||
XM_008761838.1 | XP_008760060.1 | 24 | 5932 | |||
XM_008761839.1 | XP_008760061.1 | 24 | 6218 | |||
XM_008761840.1 | XP_008760062.1 | 22 | 6850 | |||
GenBank mRNA | AB098072.1 | - | 26 | 5599 |