Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SORBS2 SIRNA Assays ›
Gene Symbol:
SORBS2
Gene Name:
sorbin and SH3 domain containing 2
Gene Aliases:
ARGBP2, PRO0618
Chromosome Location:
Chr.4: 185585444 - 185956716 on Build GRCh38
UniGene ID:
Hs.481342
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s16087
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SORBS2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145670.1
NP_001139142.1
18
2496
NM_001145671.2
NP_001139143.1
20
2285
NM_001145672.1
NP_001139144.1
18
1923
NM_001145673.1
NP_001139145.1
20
2809
NM_001145674.1
NP_001139146.1
12
3096
NM_001145675.1
NP_001139147.1
12
1560
NM_001270771.1
NP_001257700.1
21
4131
NM_003603.6
NP_003594.3
20
2524
NM_021069.4
NP_066547.1
18
3911
XM_005263307.1
XP_005263364.1
22
3403
XM_005263308.1
XP_005263365.1
21
2761
XM_005263310.1
XP_005263367.1
21
2617
XM_005263311.1
XP_005263368.1
20
2602
XM_005263312.1
XP_005263369.1
20
2599
XM_005263313.1
XP_005263370.1
18
2500
XM_006714388.1
XP_006714451.1
21
2692
XM_006714390.1
XP_006714453.1
19
2509
XM_017008741.1
XP_016864230.1
24
3961
XM_017008748.1
XP_016864237.1
21
3661
XM_017008750.1
XP_016864239.1
24
3961
XM_017008751.1
XP_016864240.1
23
3877
XM_017008752.1
XP_016864241.1
23
3802
XM_017008753.1
XP_016864242.1
21
3661
XM_017008754.1
XP_016864243.1
20
3616
XM_017008755.1
XP_016864244.1
18
2626
XM_017008756.1
XP_016864245.1
17
2329
XM_017008757.1
XP_016864246.1
13
1731
XM_017008758.1
XP_016864247.1
21
2795
XM_017008759.1
XP_016864248.1
18
3222
XM_017008760.1
XP_016864249.1
20
2668
XM_017008762.1
XP_016864251.1
19
2545
XM_017008763.1
XP_016864252.1
18
2495
XM_017008764.1
XP_016864253.1
19
2476
XM_017008765.1
XP_016864254.1
20
2779
XM_017008766.1
XP_016864255.1
18
2452
XM_017008767.1
XP_016864256.1
19
2755
XM_017008768.1
XP_016864257.1
18
2673
XM_017008769.1
XP_016864258.1
18
2431
XM_017008770.1
XP_016864259.1
17
2407
XM_017008771.1
XP_016864260.1
19
2549
XM_017008772.1
XP_016864261.1
18
2383
XM_017008773.1
XP_016864262.1
19
2686
XM_017008774.1
XP_016864263.1
14
2176
XM_017008775.1
XP_016864264.1
23
3916
GenBank mRNA
AB018320.1
-
18
3563
AF049884.1
-
20
2488
AF049885.1
-
19
1928
AK056628.1
-
21
3749
AK056758.1
-
13
1731
AK225327.1
-
18
2462
AK225812.1
-
20
2809
AK293400.1
-
18
1923
AK296461.1
-
12
2873
AK304653.1
-
12
1296
BC011883.1
-
20
2252
3677070
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
actin filament organization
cell adhesion
biological_process
cell migration
cell growth involved in cardiac muscle cell development
structural constituent of cytoskeleton
protein binding
cytoskeletal adaptor activity
structural constituent of muscle
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-v5ndd:80/100.66.76.213:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline