Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc30a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029918.1 | NP_001025089.1 | 1 | 614 | ||
GenBank mRNA | BC098051.1 | - | 1 | 614 | 7457462 |