Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107735.1 | NP_001101205.1 | 26 | 3329 | ||
XM_008761900.1 | XP_008760122.1 | 25 | 2847 | |||
XM_008761901.1 | XP_008760123.1 | 24 | 2823 | |||
XM_008761902.1 | XP_008760124.1 | 24 | 2772 | |||
XM_008761903.1 | XP_008760125.1 | 24 | 2754 | |||
XM_008761904.1 | XP_008760126.1 | 23 | 2730 | |||
XM_008761905.1 | XP_008760127.1 | 23 | 2679 | |||
XM_008761906.1 | XP_008760128.1 | 22 | 2655 |