Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smtnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191739.1 | NP_001178668.1 | 1 | 247 | ||
XM_006234464.2 | XP_006234526.1 | 2 | 370 | |||
XM_008761988.1 | XP_008760210.1 | 2 | 337 | |||
GenBank mRNA | FQ216695.1 | - | 386 | |||
FQ217513.1 | - | 2 | 331 |