Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181672.2 | NP_858058.1 | 8 | 1282 | ||
NM_181673.2 | NP_858059.1 | 8 | 1252 | |||
XM_017029907.1 | XP_016885396.1 | 6 | 811 | |||
XM_017029908.1 | XP_016885397.1 | 6 | 4284 | |||
GenBank mRNA | AF223393.1 | - | 4 | 919 | ||
AK313530.1 | - | 4 | 1198 | |||
AL050366.1 | - | 3 | 2509 | |||
AL833085.2 | - | 8 | 1262 | |||
BC014434.1 | - | 8 | 1183 | 4865031 | ||
BC038180.1 | - | 8 | 1215 | 5017795 | ||
BX537844.1 | - | 8 | 1240 | |||
U77413.1 | - | 4 | 931 |