Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107747.2 | NP_001101217.2 | 10 | 1000 | ||
XM_006234551.1 | XP_006234613.1 | 11 | 1037 | |||
XM_006234552.2 | XP_006234614.1 | 11 | 1067 | |||
XM_006234553.2 | XP_006234615.1 | 11 | 1037 | |||
XM_006234554.1 | XP_006234616.1 | 12 | 1079 | |||
XM_006234556.1 | XP_006234618.1 | 11 | 1014 | |||
XM_006234557.2 | XP_006234619.1 | 11 | 1044 | |||
XM_006234558.2 | XP_006234620.1 | 10 | 985 | |||
XM_006234559.2 | XP_006234621.1 | 8 | 2423 | |||
XM_006234560.2 | XP_006234622.1 | 10 | 1026 | |||
GenBank mRNA | DN932805.1 | - | 2 | 160 | ||
EV768858.1 | - | 6 | 471 | |||
FQ224002.1 | - | 10 | 960 |