Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Accs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267534.1 | NP_001254463.1 | 14 | 1386 | ||
XM_006234577.2 | XP_006234639.1 | 14 | 1391 | |||
XM_006234579.2 | XP_006234641.1 | 15 | 1463 | |||
XM_006234580.2 | XP_006234642.1 | 14 | 1480 | |||
XM_008762028.1 | XP_008760250.1 | 14 | 1519 | |||
GenBank mRNA | BC083866.1 | - | 14 | 1386 |