Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nusap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107762.1 | NP_001101232.1 | 6 | 914 | ||
XM_006234768.2 | XP_006234830.1 | 6 | 745 | |||
XM_006234769.2 | XP_006234831.1 | 6 | 745 | |||
XM_006234770.2 | XP_006234832.1 | 5 | 598 | |||
XM_008762101.1 | XP_008760323.1 | 5 | 655 | |||
XM_008762102.1 | XP_008760324.1 | 5 | 655 | |||
GenBank mRNA | BC161947.1 | - | 6 | 690 | 9025380 |