Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nusap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107762.1 | NP_001101232.1 | 7, 8 | 1044 | ||
XM_006234768.2 | XP_006234830.1 | 7, 8 | 878 | |||
XM_006234769.2 | XP_006234831.1 | 7, 8 | 875 | |||
XM_006234770.2 | XP_006234832.1 | 6, 7 | 731 | |||
XM_008762101.1 | XP_008760323.1 | 6, 7 | 788 | |||
XM_008762102.1 | XP_008760324.1 | 6, 7 | 785 | |||
GenBank mRNA | BC161947.1 | - | 7, 8 | 823 | 9025380 |