Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OFD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003611.2 | NP_003602.1 | 11 | 1441 | ||
XM_005274604.2 | XP_005274661.1 | 10 | 1321 | |||
XM_005274606.3 | XP_005274663.1 | 12 | 1583 | |||
XM_011545592.2 | XP_011543894.1 | 12 | 1412 | |||
XM_011545594.2 | XP_011543896.1 | 10 | 862 | |||
XM_011545595.1 | XP_011543897.1 | 10 | 872 | |||
XM_011545597.1 | XP_011543899.1 | 12 | 1497 | |||
XM_017029909.1 | XP_016885398.1 | 11 | 1017 | |||
XM_017029910.1 | XP_016885399.1 | 12 | 1225 | |||
XM_017029911.1 | XP_016885400.1 | 3 | 262 | |||
GenBank mRNA | AK225847.1 | - | 10 | 1251 | ||
AK290354.1 | - | 11 | 1426 | |||
AK297104.1 | - | 3 | 221 | |||
AK311067.1 | - | 4 | 371 | |||
BC096344.3 | - | 10 | 1136 | 40008704 | ||
BC096345.1 | - | 11 | 1262 | |||
GU727634.1 | - | 10 | 1082 | |||
Y15164.1 | - | 11 | 1393 | |||
Y16355.1 | - | 11 | 2057 |