Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100910478 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749549.3 | XP_003749597.1 | 4 | 357 | ||
XM_006234812.1 | XP_006234874.1 | 4 | 591 | |||
XM_006234813.2 | XP_006234875.1 | 3 | 303 |
Gene Symbol: Lrrc57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012354.2 | NP_001012354.1 | 591 | |||
XM_006234777.2 | XP_006234839.1 | 357 | ||||
XM_006234778.1 | XP_006234840.1 | 372 | ||||
XM_006234779.2 | XP_006234841.1 | 387 | ||||
XM_006234780.2 | XP_006234842.1 | 408 | ||||
GenBank mRNA | BC089966.1 | - | 408 | 7314989 | ||
CV111644.1 | - | 591 |