Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234874.2 | XP_006234936.1 | 14 | 2418 | ||
XM_006234875.2 | XP_006234937.1 | 13 | 2325 | |||
XM_006234876.2 | XP_006234938.1 | 13 | 1863 | |||
XM_006234877.1 | XP_006234939.1 | 13 | 1856 | |||
XM_008762207.1 | XP_008760429.1 | 10 | 1535 | |||
XM_008762208.1 | XP_008760430.1 | 11 | 1457 | |||
XM_008762209.1 | XP_008760431.1 | 10 | 1284 | |||
XM_008762210.1 | XP_008760432.1 | 10 | 1222 | |||
XM_230512.7 | XP_230512.4 | 13 | 1894 |