Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Duoxa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107767.1 | NP_001101237.1 | 4, 5 | 573 | ||
XM_006234857.1 | XP_006234919.1 | 5, 6 | 461 |