Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema6d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107768.1 | NP_001101238.1 | 12 | 1818 | ||
XM_008762171.1 | XP_008760393.1 | 12 | 5254 | |||
XM_008762172.1 | XP_008760394.1 | 12 | 5254 | |||
XM_008762173.1 | XP_008760395.1 | 12 | 5254 | |||
XM_008762174.1 | XP_008760396.1 | 12 | 5254 | |||
XM_008762175.1 | XP_008760397.1 | 12 | 5254 |