Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100978.1 | NP_001094448.1 | 2, 3 | 444 | ||
GenBank mRNA | BC131850.1 | - | 2, 3 | 444 | 7125680 | |
FQ230469.1 | - | 2, 3 | 418 |