Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107774.1 | NP_001101244.1 | 7, 8 | 552 | ||
GenBank mRNA | FQ230286.1 | - | 7, 8 | 515 | ||
FQ231907.1 | - | 7, 8 | 515 |