Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254748.1 | NP_001241677.1 | 3 | 332 | ||
NM_001254749.1 | NP_001241678.1 | 2 | 447 | |||
NM_003617.3 | NP_003608.1 | 2 | 447 | |||
GenBank mRNA | AB008109.1 | - | 2 | 190 | ||
AF030108.1 | - | 2 | 185 | |||
AF159570.1 | - | 2 | 153 | |||
AF493929.1 | - | 2 | 109 | |||
AK315357.1 | - | 2 | 190 | |||
AU130764.1 | - | 2 | 356 | |||
BC030059.1 | - | 2 | 172 | 4098248 | ||
BP292867.1 | - | 3 | 292 | |||
BU172751.1 | - | 2 | 177 | |||
BX537427.1 | - | 2 | 191 | |||
DA699380.1 | - | 2 | 425 | |||
DA731601.1 | - | 2 | 448 |