Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sel1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014049.1 | NP_001014071.1 | 8 | 926 | ||
GenBank mRNA | BC083893.1 | - | 8 | 926 | 7132857 |