Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kiz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235181.2 | XP_006235243.1 | 5 | 1016 | ||
XM_006235182.2 | XP_006235244.1 | 5 | 1016 | |||
XM_008762291.1 | XP_008760513.1 | 5 | 1016 | |||
XM_008762292.1 | XP_008760514.1 | 2 | 787 |