Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam110a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014050.2 | NP_001014072.2 | 2 | 532 | ||
NM_001244768.1 | NP_001231697.1 | 2 | 413 | |||
NM_001244769.1 | NP_001231698.1 | 2 | 485 | |||
NM_001244770.1 | NP_001231699.1 | 2 | 332 | |||
XM_006235231.2 | XP_006235293.1 | 2 | 785 | |||
XM_006235234.2 | XP_006235296.1 | 3 | 520 | |||
GenBank mRNA | BC079251.1 | - | 2 | 532 | 7133086 | |
BU670818.1 | - | 2 | 331 | |||
CV110036.1 | - | 2 | 413 | |||
CV113127.1 | - | 2 | 485 | |||
DY318143.1 | - | 1 | 213 | |||
JF912497.1 | - | 1 | 143 |