Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rprd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098727.1 | NP_001092197.1 | 6 | 1073 | ||
XM_006235412.1 | XP_006235474.1 | 6 | 1076 | |||
GenBank mRNA | BC168699.1 | - | 6 | 1078 | 8362231 | |
CK365545.1 | - | 4 | 321 |