Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfatc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107805.1 | NP_001101275.1 | 2 | 715 | ||
XM_006235653.1 | XP_006235715.1 | 2 | 709 | |||
XM_008762507.1 | XP_008760729.1 | 2 | 709 |