Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RANBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300865.1 | NP_001287794.1 | 9, 10 | 919 | ||
NM_003624.2 | NP_003615.2 | 10, 11 | 1123 | |||
NM_007320.2 | NP_015559.2 | 9, 10 | 934 | |||
NM_007322.2 | NP_015561.1 | 10, 11 | 1138 | |||
XM_006722928.1 | XP_006722991.1 | 11, 12 | 1125 | |||
XM_006722929.1 | XP_006722992.1 | 9, 10 | 761 | |||
XM_011528391.1 | XP_011526693.1 | 9, 10 | 884 | |||
XM_011528392.1 | XP_011526694.1 | 10, 11 | 1018 | |||
XM_011528393.1 | XP_011526695.1 | 9, 10 | 729 | |||
XM_011528394.1 | XP_011526696.1 | 7, 8 | 590 | |||
XM_017027405.1 | XP_016882894.1 | 6, 7 | 580 | |||
XM_017027406.1 | XP_016882895.1 | 6, 7 | 552 | |||
GenBank mRNA | AK025300.1 | - | 10, 11 | 974 | ||
AK125183.1 | - | 5, 6 | 1473 | |||
AK222813.1 | - | 9, 10 | 718 | |||
AK222990.1 | - | 10, 11 | 922 | |||
AK299260.1 | - | 6, 7 | 585 | |||
AK299856.1 | - | 9, 10 | 867 | |||
AK301942.1 | - | 9, 10 | 703 | |||
AK308205.1 | - | 10, 11 | 850 | |||
AK314343.1 | - | 9, 10 | 765 | |||
AL050149.1 | - | 10, 11 | 921 | |||
BC004349.1 | - | 9, 10 | 716 | 3350317 | ||
Y08697.1 | - | 8, 9 | 699 | |||
Y08698.1 | - | 10, 11 | 916 |