Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il36rn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107814.1 | NP_001101284.1 | 4 | 367 | ||
XM_008761586.1 | XP_008759808.1 | 2 | 1018 | |||
XM_008761587.1 | XP_008759809.1 | 3 | 661 |