Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134881.1 | NP_001128353.1 | 10 | 2288 | ||
XM_006233591.2 | XP_006233653.1 | 8 | 2371 | |||
XM_006233592.2 | XP_006233654.1 | 8 | 2156 | |||
XM_006233593.2 | XP_006233655.1 | 8 | 2362 |