Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012094.1 | NP_001012094.1 | 4 | 1562 | ||
XM_006233973.2 | XP_006234035.1 | 3 | 1542 | |||
XM_006233974.2 | XP_006234036.1 | 2 | 1481 | |||
XM_008761749.1 | XP_008759971.1 | 2 | 2589 | |||
GenBank mRNA | BC087042.1 | - | 4 | 1562 | 7107163 |