Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109666.1 | NP_001103136.1 | 9 | 1387 | ||
XM_006235929.2 | XP_006235991.1 | 10 | 1357 | |||
XM_006235930.2 | XP_006235992.1 | 10 | 1519 | |||
XM_006235931.2 | XP_006235993.1 | 10 | 1354 | |||
XM_006235932.2 | XP_006235994.1 | 10 | 1348 | |||
XM_006235933.2 | XP_006235995.1 | 10 | 1345 | |||
GenBank mRNA | BC129119.1 | - | 9 | 1254 | 7115823 |