Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256574.1 | NP_001243503.1 | 3 | 2656 | ||
NM_001256575.1 | NP_001243504.1 | 3 | 2644 | |||
NM_001256576.1 | NP_001243505.1 | 2 | 2383 | |||
NM_003633.3 | NP_003624.1 | 2 | 2519 | |||
XM_011543696.2 | XP_011541998.1 | 3 | 2727 | |||
XM_011543697.2 | XP_011541999.1 | 3 | 2573 | |||
GenBank mRNA | AF005381.1 | - | 3 | 1544 | ||
AF010314.1 | - | 2 | 1824 | |||
AF059611.1 | - | 2 | 1787 | |||
AK096578.1 | - | 2 | 2519 | |||
AK295128.1 | - | 2 | 1688 | |||
AK299624.1 | - | 2 | 713 | |||
AK312608.1 | - | 2 | 1821 | |||
AY049781.1 | - | 1 | 1392 | |||
BC000418.2 | - | 2 | 1789 | 2964376 | ||
BT007392.1 | - | 1 | 1388 |