Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtpbp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100815.1 | NP_001094285.1 | 4, 5 | 538 | ||
GenBank mRNA | CV103632.1 | - | 2, 3 | 204 |