Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173511.1 | NP_001166982.1 | 16, 17 | 1444 | ||
XM_006236037.2 | XP_006236099.1 | 16, 17 | 1475 | |||
XM_008762704.1 | XP_008760926.1 | 16, 17 | 1478 | |||
GenBank mRNA | BC091288.1 | - | 1, 2 | 39 |