Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107849.2 | NP_001101319.2 | 4 | 1075 | ||
XM_006257073.2 | XP_006257135.1 | 5 | 2583 | |||
XM_006257074.2 | XP_006257136.1 | 6 | 1315 | |||
XM_006257075.2 | XP_006257137.1 | 5 | 1199 |