Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIPSNAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202502.1 | NP_001189431.1 | 2, 3 | 181 | ||
NM_003634.3 | NP_003625.2 | 2, 3 | 482 | |||
GenBank mRNA | AJ001258.1 | - | 2, 3 | 481 | ||
AK098367.1 | - | 2, 3 | 236 | |||
AK290953.1 | - | 2, 3 | 235 | |||
AK298816.1 | - | 2, 3 | 183 | |||
AK314407.1 | - | 2, 3 | 253 | |||
BC002371.2 | - | 2, 3 | 232 | 2822369 | ||
BC006473.1 | - | 2, 3 | 232 | 2822369 | ||
BC018869.2 | - | 1, 2 | 124 | |||
DA133702.1 | - | 2, 3 | 300 |