Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIPF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198828.1 | NP_001185757.1 | 5 | 485 | ||
NM_001198829.1 | NP_001185758.1 | 6 | 665 | |||
NM_001198830.1 | NP_001185759.1 | 6 | 566 | |||
NM_004190.3 | NP_004181.1 | 5 | 584 | |||
XM_011540311.1 | XP_011538613.1 | 5 | 526 | |||
GenBank mRNA | A01046.1 | - | 5 | 516 | ||
AK223468.1 | - | 5 | 529 | |||
AK301310.1 | - | 5 | 434 | |||
AK301320.1 | - | 6 | 613 | |||
AK308224.1 | - | 5 | 532 | |||
AK312940.1 | - | 5 | 531 | |||
AL833751.1 | - | 6 | 497 | |||
BC112272.1 | - | 4 | 503 | 8327740 | ||
BC113711.1 | - | 4 | 492 | 8322763 | ||
X05997.1 | - | 5 | 514 |