Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107858.1 | NP_001101328.1 | 10 | 1824 | ||
XM_008762883.1 | XP_008761105.1 | 10 | 1819 |