Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIPF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198828.1 | NP_001185757.1 | 2, 3 | 221 | ||
NM_001198829.1 | NP_001185758.1 | 3, 4 | 302 | |||
NM_001198830.1 | NP_001185759.1 | 3, 4 | 302 | |||
NM_004190.3 | NP_004181.1 | 2, 3 | 221 | |||
XM_011540311.1 | XP_011538613.1 | 2, 3 | 163 | |||
GenBank mRNA | A01046.1 | - | 2, 3 | 153 | ||
AK223468.1 | - | 2, 3 | 166 | |||
AK301310.1 | - | 2, 3 | 170 | |||
AK301320.1 | - | 3, 4 | 250 | |||
AK308224.1 | - | 2, 3 | 169 | |||
AK310128.1 | - | 2, 3 | 169 | |||
AK312940.1 | - | 2, 3 | 168 | |||
AL833751.1 | - | 3, 4 | 233 | |||
BC112272.1 | - | 1, 2 | 140 | 8327740 | ||
BC113711.1 | - | 1, 2 | 129 | 8322763 | ||
BP341513.1 | - | 2, 3 | 221 | |||
DA967766.1 | - | 2, 3 | 275 | |||
X05997.1 | - | 2, 3 | 151 |