Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarcad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107864.2 | NP_001101334.2 | 8 | 1067 | ||
XM_006236599.1 | XP_006236661.1 | 9 | 1150 | |||
XM_006236600.2 | XP_006236662.1 | 9 | 1216 | |||
XM_006236601.2 | XP_006236663.1 | 8 | 2306 | |||
XM_008762979.1 | XP_008761201.1 | 8 | 3081 |