Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Immt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034928.1 | NP_001030100.1 | 13, 14 | 1601 | ||
XM_008762981.1 | XP_008761203.1 | 14, 15 | 1831 | |||
XM_008762982.1 | XP_008761204.1 | 13, 14 | 1816 | |||
XM_008762983.1 | XP_008761205.1 | 14, 15 | 1797 | |||
XM_008762984.1 | XP_008761206.1 | 14, 15 | 1794 | |||
XM_008762985.1 | XP_008761207.1 | 13, 14 | 1782 | |||
XM_008762986.1 | XP_008761208.1 | 13, 14 | 1733 | |||
XM_008762987.1 | XP_008761209.1 | 13, 14 | 1699 | |||
XM_008762988.1 | XP_008761210.1 | 13, 14 | 1696 | |||
XM_008762989.1 | XP_008761211.1 | 12, 13 | 1684 | |||
XM_008762990.1 | XP_008761212.1 | 12, 13 | 1606 | |||
GenBank mRNA | BC105841.1 | - | 13, 14 | 1601 | 7454026 | |
FQ217458.1 | - | 12, 13 | 1528 |