Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303040.1 | NP_001289969.1 | 22 | 2870 | ||
NM_001303041.1 | NP_001289970.1 | 21 | 2758 | |||
NM_003637.4 | NP_003628.2 | 25 | 3187 | |||
XM_005277436.3 | XP_005277493.1 | 24 | 2938 | |||
XM_011510083.2 | XP_011508385.1 | 21 | 2618 | |||
XM_011510084.1 | XP_011508386.1 | 14 | 1615 | |||
XM_017002622.1 | XP_016858111.1 | 23 | 3144 | |||
XM_017002623.1 | XP_016858112.1 | 23 | 3141 | |||
XM_017002624.1 | XP_016858113.1 | 23 | 3144 | |||
XM_017002626.1 | XP_016858115.1 | 21 | 2901 | |||
XM_017002628.1 | XP_016858117.1 | 19 | 2594 | |||
GenBank mRNA | AF074015.1 | - | 25 | 3033 | ||
AF112345.1 | - | 25 | 3088 | |||
AK304155.1 | - | 22 | 2726 | |||
AK304183.1 | - | 24 | 3017 | |||
AK314255.1 | - | 25 | 3036 | |||
BC140831.1 | - | 21 | 2633 | 9021699 | ||
BC144637.1 | - | 21 | 2633 | 9053168 |