Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010956.2 | NP_001010956.1 | 4, 5 | 787 | ||
NM_001033931.1 | NP_001029103.1 | 4, 5 | 787 | |||
XM_006236832.2 | XP_006236894.1 | 4, 5 | 802 | |||
GenBank mRNA | AY862307.1 | - | 4, 5 | 614 | ||
AY862308.1 | - | 4, 5 | 582 |