Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Necap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029919.2 | NP_001025090.1 | 2 | 185 | ||
XM_006237305.1 | XP_006237367.1 | 3 | 246 | |||
GenBank mRNA | BC097496.1 | - | 2 | 185 | 7461823 |