Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017380.1 | NP_001017380.1 | 14 | 2440 | ||
XM_008772408.1 | XP_008770630.1 | 14 | 3689 | |||
GenBank mRNA | BC082001.1 | - | 14 | 2440 |
Gene Symbol: Cyld-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763560.1 | XP_008761782.1 | 1 | 2453 |