Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237894.2 | XP_006237956.1 | 2 | 4578 | ||
XM_006237895.2 | XP_006237957.1 | 2 | 3715 | |||
XM_006237897.2 | XP_006237959.1 | 3 | 3201 | |||
XM_006237898.2 | XP_006237960.1 | 3 | 2608 | |||
XM_006237899.2 | XP_006237961.1 | 2 | 4015 | |||
XM_008763579.1 | XP_008761801.1 | 2 | 3122 |