Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100710.1 | NP_001094180.1 | 2 | 1208 | ||
GenBank mRNA | BC105891.1 | - | 1 | 527 |