Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003654.5 | NP_003645.1 | 4 | 1447 | ||
XM_006718356.3 | XP_006718419.1 | 4 | 1412 | |||
XM_017018459.1 | XP_016873948.1 | 4 | 1453 | |||
GenBank mRNA | AB003791.1 | - | 4 | 1108 | ||
AF090137.1 | - | 1 | 746 | |||
BC022567.1 | - | 4 | 1413 | 4814499 | ||
BC028235.1 | - | 4 | 1081 | 5245338 | ||
U65637.1 | - | 2 | 866 |