Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcaf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107941.1 | NP_001101411.1 | 2 | 1645 | ||
XM_006257515.2 | XP_006257577.1 | 2 | 1845 | |||
XM_008773519.1 | XP_008771741.1 | 2 | 1845 |