Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thoc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191756.1 | NP_001178685.1 | 23 | 2682 | ||
XM_006257504.2 | XP_006257566.1 | 23 | 2693 | |||
XM_006257505.2 | XP_006257567.1 | 23 | 2693 | |||
GenBank mRNA | FQ232536.1 | - | 2 | 305 |