Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004041.4 | NP_004032.2 | 15 | 1353 | ||
NM_020251.3 | NP_064647.1 | 14 | 1329 | |||
XM_011545034.2 | XP_011543336.1 | 16 | 1237 | |||
XM_011545035.2 | XP_011543337.1 | 15 | 1213 | |||
XM_017017750.1 | XP_016873239.1 | 16 | 1237 | |||
XM_017017751.1 | XP_016873240.1 | 15 | 1213 | |||
XM_017017752.1 | XP_016873241.1 | 15 | 1193 | |||
XM_017017753.1 | XP_016873242.1 | 15 | 1257 | |||
XM_017017754.1 | XP_016873243.1 | 14 | 1169 | |||
GenBank mRNA | AF084040.1 | - | 15 | 1129 | ||
AF084940.1 | - | 14 | 1105 | |||
AK289718.1 | - | 15 | 1183 | |||
BC003636.2 | - | 15 | 1206 | 3604829 | ||
FJ348262.1 | - | 14 | 1105 | |||
L04685.1 | - | 15 | 1129 |